Dextrin, yellow
Glass bottle for solids, 1 kg
Katalognr. ROTH6777.1
Marketsource
Anbieter: Roth Carl
Direkt zu
Product Details & Documents
Synonyme: Dextrin
- CAS Nummer:9004-53-9
- Formel:(C₆H₁₂O₆)n
- MDL-Nummer:MFCD00081554
- Molekulargewicht:180.16 g/mol
- Lagertemperatur:Ambient
- EINECS:232-675-4
Technische Daten
Technische Daten
- Pack type:Glass bottle for solids
- Pk:1 kg



