Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Sodium hydroxide

Sodium hydroxide

Glass bottle for solids, 500 g
Catalog # ROTH6771.3
Corrosive
Marketsource
Supplier:   Roth Carl
Sodium hydroxide
Catalog # ROTH6771.3
Supplier:  Roth Carl
Marketsource
CAS Number:  1310-73-2
Sodium hydroxide

Glass bottle for solids, 500 g

CAS Number:  1310-73-2
Price
SFr. 32.30
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,08627
  • Boiling Pt:1390 °C (1013 hPa)
  • Melting Pt:323 °C
  • Density:2.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1823
  • ADR:8,II
  • REACH:01-2119457892-27
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.