Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Sodium hydroxide

Sodium hydroxide

Glass bottle for solids, 1 kg
Catalog # ROTH6771.1
Corrosive
Marketsource
Supplier:   Roth Carl
Sodium hydroxide
Catalog # ROTH6771.1
Supplier:  Roth Carl
Marketsource
CAS Number:  1310-73-2
Sodium hydroxide

Glass bottle for solids, 1 kg

CAS Number:  1310-73-2
Price
SFr. 53.90
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,08627
  • Boiling Pt:1390 °C (1013 hPa)
  • Melting Pt:323 °C
  • Density:2.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1823
  • ADR:8,II
  • REACH:01-2119457892-27
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..