Chloroform
Glass bottle, 100 mL
Catalog # ROTH6340.4
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
- CAS Number:67-66-3
- Formula:CHCl₃
- MDL Number:MFCD00000826
- Molecular Weight:119.38 g/mol
- Merck Index:13,02160
- Boiling Pt:61.7 °C (1013 hPa)
- Melting Pt:–63 °C
- Density:1.4832 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-663-8
- UN:1888
- ADR:6.1,III
- REACH:05-2114614906-45
Specifications
Specifications
- Pack type:Glass bottle
- Pk:100 mL



