Propionic acid
Glass bottle, 1 L
Catalog # ROTH6026.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Propanoic acid
- CAS Number:79-09-4
- Formula:C₃H₆O₂
- MDL Number:MFCD00002756
- Molecular Weight:74.08 g/mol
- Merck Index:13,07917
- Boiling Pt:141 °C (1013 hPa)
- Melting Pt:–21 °C
- Density:0.99 g/cm³ (20 °C)
- Flash Pt:49 °C
- Storage Temperature:Ambient
- EINECS:201-176-3
- UN:3463
- ADR:8,II
- REACH:01-2119486971-24
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L



