Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Formic acid 85%

Formic acid 85%

10 L
Catalog # ROTH5355.3
Corrosive
Marketsource
Supplier:   Roth Carl
Formic acid 85%
Catalog # ROTH5355.3
Supplier:  Roth Carl
Marketsource
CAS Number:  64-18-6
Formic acid 85%
Formic acid 85%

10 L

CAS Number:  64-18-6
Price
SFr. 141.50
Log in for availability
1 * 10 L
Product Details & Documents
Synonyms:  Aminic Acid, Methanoic acid
  • CAS Number:64-18-6
  • Formula:HCO₂H
  • MDL Number:MFCD00003297
  • Molecular Weight:46.03 g/mol
  • Merck Index:13,04265
  • ADR:8,II
  •  :Density: 1.22 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:200-579-1
  • UN:1779
Specifications

Specifications

  • Pk:
    10 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.