Order Entry
Switzerland
Orders LinkContactUsLinkComponent
D-(+)-Galactose

D-(+)-Galactose

Glass bottle for solids, 100 g
Catalog # ROTH4987.1
Marketsource
Supplier:   Roth Carl
D-(+)-Galactose
Catalog # ROTH4987.1
Supplier:  Roth Carl
Marketsource
CAS Number:  59-23-4
D-(+)-Galactose
D-(+)-Galactose

Glass bottle for solids, 100 g

CAS Number:  59-23-4
Price
SFr. 39.50
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
  • CAS Number:59-23-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00151230
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04356
  • Boiling Pt:527 °C (1013 hPa)
  • Melting Pt:163…169 °C
  • Density:1.5 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-416-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications