Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Methanol

Methanol

Glass bottle, 1 L
Catalog # ROTH4627.1
Flammable
Toxic
Health
Marketsource
Supplier:   Roth Carl
Methanol
Catalog # ROTH4627.1
Supplier:  Roth Carl
Marketsource
CAS Number:  67-56-1
Methanol
Methanol

Glass bottle, 1 L

CAS Number:  67-56-1
Price
39.50 SFr.
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
  • CAS Number:67-56-1
  • Formula:CH₃OH
  • MDL Number:MFCD00004595
  • Molecular Weight:32.04 g/mol
  • Merck Index:13,05984
  • Boiling Pt:64.6 °C (1013 hPa)
  • Melting Pt:–98 °C
  • Density:0.7918 g/cm³ (20 °C)
  • Flash Pt:11 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-659-6
  • UN:1230
  • ADR:3,II
  • REACH:01-2119433307-44
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.