Order Entry
Switzerland
Orders LinkContactUsLinkComponent
1-Octanol

1-Octanol

Glass bottle, 100 mL
Catalog # ROTH4439.1
Irritant
Marketsource
Supplier:   Roth Carl
1-Octanol
Catalog # ROTH4439.1
Supplier:  Roth Carl
Marketsource
CAS Number:  111-87-5
1-Octanol
1-Octanol

Glass bottle, 100 mL

CAS Number:  111-87-5
Price
27.00 SFr.
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  n-Octanol
  • CAS Number:111-87-5
  • Formula:H₃C(CH₂)₆CH₂OH
  • MDL Number:MFCD00002988
  • Molecular Weight:130.23 g/mol
  • Merck Index:13,06784
  • Boiling Pt:195 °C (1013 hPa)
  • Melting Pt:–16 °C
  • Density:0.8246 g/cm³ (20 °C)
  • Flash Pt:81 °C
  • Storage Temperature:Ambient
  • EINECS:203-917-6
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications