Order Entry
Switzerland
Orders LinkContactUsLinkComponent
2-Mercaptoethanol

2-Mercaptoethanol

Glass bottle, 1 L
Catalog # ROTH4227.2
Toxic
Corrosive
Environment
Marketsource
Supplier:   Roth Carl
2-Mercaptoethanol
Catalog # ROTH4227.2
Supplier:  Roth Carl
Marketsource
CAS Number:  60-24-2
2-Mercaptoethanol
2-Mercaptoethanol

Glass bottle, 1 L

CAS Number:  60-24-2
Price
SFr. 75.50
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  2-Hydroxyethyl mercaptan, β-Mercaptoethanol
  • CAS Number:60-24-2
  • Formula:HSCH₂CH₂OH
  • MDL Number:MFCD00004890
  • Molecular Weight:78.14 g/mol
  • Merck Index:13,05894
  • Boiling Pt:157 °C (1013 hPa)
  • Melting Pt:–100 °C
  • Density:1.12 g/cm³ (20 °C)
  • Flash Pt:68 °C
  • Storage Temperature:Ambient
  • EINECS:200-464-6
  • UN:2966
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.