Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

1-Hexanol

Glass bottle, 1 L
Catalog # ROTH3847.1
Irritant
Marketsource
Supplier:   Roth Carl
1-Hexanol
Catalog # ROTH3847.1
Supplier:  Roth Carl
Marketsource
CAS Number:  111-27-3
1-Hexanol
1-Hexanol

Glass bottle, 1 L

CAS Number:  111-27-3
Price
68.30 SFr.
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Hexyl alcohol, Pentyl carbinol, 1-Hexyl alcohol, 1-Hydroxy hexane
  • CAS Number:111-27-3
  • Formula:H₃C(CH₂)₄CH₂OH
  • MDL Number:MFCD00002982
  • Molecular Weight:102.18 g/mol
  • Merck Index:13,04715
  • Boiling Pt:156 °C (1013 hPa)
  • Melting Pt:–45 °C
  • Density:0.8322 g/cm³ (20 °C)
  • Flash Pt:60 °C
  • Storage Temperature:Ambient
  • EINECS:203-852-3
  • UN:2282
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications