Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Chloroform

Chloroform

25 L
Catalog # ROTH3313.5
Toxic
Health
Marketsource
Supplier:   Roth Carl
Chloroform
Catalog # ROTH3313.5
Supplier:  Roth Carl
Marketsource
CAS Number:  67-66-3
Chloroform
Chloroform

25 L

CAS Number:  67-66-3
Price
1 242.00 SFr.
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
  • CAS Number:67-66-3
  • Formula:CHCl₃
  • MDL Number:MFCD00000826
  • Molecular Weight:119.38 g/mol
  • Merck Index:13,02160
  • Boiling Pt:61.7 °C (1013 hPa)
  • Melting Pt:–63 °C
  • Density:1.4832 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-663-8
  • UN:1888
  • ADR:6.1,III
  • REACH:05-2114614906-45
Specifications

Specifications

  • Pk:
    25 L

Specifications