Chloroform
Glass bottle, 1 L
Catalog # ROTH3313.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
- CAS Number:67-66-3
- Formula:CHCl₃
- MDL Number:MFCD00000826
- Molecular Weight:119.38 g/mol
- Merck Index:13,02160
- Boiling Pt:61.7 °C (1013 hPa)
- Melting Pt:–63 °C
- Density:1.4832 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-663-8
- UN:1888
- ADR:6.1,III
- REACH:05-2114614906-45
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



