Order Entry
Switzerland
Orders LinkContactUsLinkComponent
L(+)-Lithium lactate ≥97%

L(+)-Lithium lactate ≥97%

Glass bottle for solids, 50 g
Catalog # A11818.18
L(+)-Lithium lactate ≥97%
Catalog # A11818.18
CAS Number:  27848-80-2
L(+)-Lithium lactate ≥97%
L(+)-Lithium lactate ≥97%

Glass bottle for solids, 50 g

CAS Number:  27848-80-2
Price
92.90 SFr.
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 97% CAS No.: 27848-80-2 Molecular Formula: C3H5LiO3 Formula Weight: 96.01 Melting Point: 253-255° Beilstein Reference: 3568332

Synonyms:  Lithium (S)-2-hydroxypropanoate, (S)-(+)-Lactic acid lithium salt, (S)-2-Hydroxypropionic acid Lithium salt, Lithium (S)-2-Hydroxypropionate, L(+)-Lactic acid lithium salt, (S)-(+)-2-Hydroxypropanoic acid lithium salt, (S)-(+)-2-Hydroxypropionic acid lithium salt, Lithium L-Lactate, L-Lactic acid Lithium salt
  • CAS Number:27848-80-2
  • Formula:C₃H₅LiO₃
  • MDL Number:MFCD00065512
  • Molecular Weight:96.01 g/mol
  • Melting Pt:253…255 °C
  • Storage Temperature:Ambient
  • EINECS:248-692-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.