Cleaning solution 500ml for410 flame photometer 1 * 500 mL
Catalog # SHEW00156184
Supplier: SHERWOOD SCIENTIFIC INSTRUMENT
Cleaning solution 500ml for410 flame photometer 1 * 500 mL
Catalog # SHEW00156184
Supplier: SHERWOOD SCIENTIFIC INSTRUMENT
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Potassium dihydrogen phosphate ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Combitips® advanced Eppendorf Quality Positive Displacement Pipette Tips
Supplier: EPPENDORF
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR® Beakers, Disposable
Supplier: VWR Collection
VWR®, Weighing Bottles, Glass
Supplier: VWR Collection