Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Clamps

Clamps

20 - 40 mm
Catalog # 241-0373
Supplier:   USBECK Laborgeräte
Clamps
Catalog # 241-0373
Clamps
Clamps

20 - 40 mm

Price
SFr. 25.00
Log in for availability
1 * 1 ST
Product Details & Documents

Zinc die-casting, nickel plated.

  • With vinyl coated jaws
  • With rod Ø 10 mm
Specifications
  • Spanning width:
    20 - 40 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.