Folding precision magnifiers
Folding double magnifier, biconvex, 30 mm
Catalog # 630-2599
Supplier: ESCHENBACH OPTIK
Jump to:
Product Details & Documents
Swivelling black metallic magnifier with loop, lens frame made from black painted metal.
- Protective case can serve as a handle
- Provide distortion-free images
- Chrome plated metal
Specifications
- Ø:30 mm
- Magnification:4×; 6×
- Description:Folding double magnifier, biconvex
- Seq:005