Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Drying tubes, U-shaped

Drying tubes, U-shaped

14/23 NS, 125 mm, 16 mm
Catalog # WITG2.114.125
Supplier:   witeg Labortechnik
Drying tubes, U-shaped
Catalog # WITG2.114.125
Supplier:  witeg Labortechnik
Drying tubes, U-shaped
Drying tubes, U-shaped

14/23 NS, 125 mm, 16 mm

Price
SFr. 94.40
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate glass, clear. Replaceable NS stopcocks. Lateral tube connectors.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
  • Ø int.:
    16 mm
  • Socket size:
    14/23 NS
  • Length:
    125 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.