Orders LinkContactUsLinkComponent
Beeswax
Glass bottle for solids, 250 g
  ROTH5830.1
Marketsource
 :   Roth Carl
Beeswax
  ROTH5830.1
 :  Roth Carl
Marketsource
 :  8012-89-3
Beeswax
Glass bottle for solids, 250 g
 :  8012-89-3
SFr. 28.20
1 * 250 g
 :  Wax, bees
  •  :8012-89-3
  •  :MFCD00132754
  •  :∼64 °C
  •  :0.95 g/cm³ (20 °C)
  •  :Ambient
  •  :232-383-7
  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g