Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Beeswax

Beeswax

Glass bottle for solids, 1 kg
Catalog # ROTH5830.2
Marketsource
Supplier:   Roth Carl
Beeswax
Catalog # ROTH5830.2
Supplier:  Roth Carl
Marketsource
CAS Number:  8012-89-3
Beeswax

Glass bottle for solids, 1 kg

CAS Number:  8012-89-3
Price
SFr. 77.90
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Wax, bees
  • CAS Number:8012-89-3
  • MDL Number:MFCD00132754
  • Melting Pt:∼64 °C
  • Density:0.95 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:232-383-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications