Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Starch (from corn)

Glass bottle for solids, 1 kg
Catalog # ROTH9444.1
Marketsource
Supplier:   Roth Carl
Starch (from corn)
Catalog # ROTH9444.1
Supplier:  Roth Carl
Marketsource
CAS Number:  9005-25-8
Starch (from corn)

Glass bottle for solids, 1 kg

CAS Number:  9005-25-8
Price
SFr. 52.70
Log in for availability
1 * 1 kg
Product Details & Documents
  • CAS Number:9005-25-8
  • Formula:(C₆H₁₀O₅)n
  • MDL Number:MFCD00082026
  • Molecular Weight:162.14 g/mol
  • Storage Temperature:Ambient
  • EINECS:232-679-6

 

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.