Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Dispenser tips, ritips®

BioClean®, 1,25 ml, +, Ritter ripette®, Eppendorf® Multipette 4780, 100/pack
Catalog # BZYM742111
Marketsource
Supplier:   BIOZYM
Dispenser tips, ritips®
Catalog # BZYM742111
Supplier:  BIOZYM
Marketsource
Dispenser tips, ritips®

BioClean®, 1,25 ml, +, Ritter ripette®, Eppendorf® Multipette 4780, 100/pack

Price
Inquire For Price
Log in for availability
1 * 100 ST
Product Details & Documents

These dispenser tips are compatible with Ritter ripette®, Eppendorf Multipette® 4780 and many other dispensing systems.

Bioclean® tips are sterile and free from DNase, RNase and pyrogens.

 : The 25 ml and 50 ml tips are supplied with adapter.

Specifications
  • Packed:
    100/pack
  • Sterile:
    +
  • Volume:
    1,25 ml
  • Seq:
    06
  • Tip style:
    BioClean®
  • For use with:
    Ritter ripette®, Eppendorf® Multipette 4780
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.