Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Goggles, V-Maxx

Goggles, V-Maxx

Neoprene strap; ventilated; anti-UV (2-1,2), anti-scratch, anti-fog, Clear acetate, Transparent
Catalog # 111-0736
Supplier:   HONEYWELL SAFETY
Goggles, V-Maxx
Catalog # 111-0736
Supplier:  HONEYWELL SAFETY
Goggles, V-Maxx
Goggles, V-Maxx

Neoprene strap; ventilated; anti-UV (2-1,2), anti-scratch, anti-fog, Clear acetate, Transparent

Price
22.80 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

Sporty design goggles with elastic strap.

  • 180° field of vision for distortion-free peripheral vision
  • Rotating strap attachments for easier use with hardhats
  • Indirect ventilation complies with splash-resistance test

Can be worn over prescription eyewear and some half mask respirators.

Weight: 93 g.

Certifications: EN 166, EN 169, EN 170, 1 F, C, 3, 4, 9

Specifications
  • Type:
    Neoprene strap; ventilated; anti-UV (2-1,2), anti-scratch, anti-fog
  • Colour:
    Transparent
  • Seq:
    06
  • Lenses:
    Clear acetate
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.