Robotic tips, Axygen®, for Beckman®
FXF-165-WB-R-S (wide bore), +, Transparent, AP-96 / SPAN-8 / MP200
Catalog # 736-0269
Jump to:
Discontinued
Product Details & Documents
Free of RNase, DNase, DNA and pyrogens.
- Transparent PP
- Sterile tips: Sterilised by gamma radiation
- Filter tips: PE filter prevents any contamination of the pipette
- Maxymum Recovery™ tips: Ultra-smooth surface prevents retention, loss of components, and denaturation of the sample
Documents
Specifications
- Tip style:FXF-165-WB-R-S (wide bore)
- Tip colour:Transparent
- For use with:AP-96 / SPAN-8 / MP200
- Filter:+