Pycnometers, unadjusted
1 ml
Katalognr. WITG3.900.001
Anbieter: witeg Labortechnik
Direkt zu
Product Details & Documents
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
- With ground-in capillary stopper
- Nominal capacity printed on the bottom
- Very good chemical and temperature resistance
Zertifizierung: DIN 12797
Technische Daten
- Volume (nom.):1 ml