Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Boxes with Snap-On Lid, Multipurpose

1000 ml, 103×103×127 mm
N° de catalogue BURK6250-1000
Fournisseur:   BURKLE
Boxes with Snap-On Lid, Multipurpose
N° de catalogue BURK6250-1000
Fournisseur:  BURKLE
Boxes with Snap-On Lid, Multipurpose
Boxes with Snap-On Lid, Multipurpose

1000 ml, 103×103×127 mm

Prix
SFr. 6.60
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

Box for storing, packing, taking samples, sampling and transport of pastes, granules, powders, soil samples and frozen goods (−40 °C up to briefly +95 °C).
PE, translucent.

  • Can be deep frozen
  • No crevices
  • Suitable for use with foodstuffs
Spécifications
  • Capacity:
    1000 ml
  • Seq:
    06
  • W×D×H:
    103×103×127 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.