Orders LinkContactUsLinkComponent
Filling solutions for combined/reference electrodes
Filling solutions for combined/reference electrodes
KS120 KCl solution, saturated with AgCl, 500 ml bottle, 500 mL
Catalog # HACHC20C310
Supplier:   Hach
Filling solutions for combined/reference electrodes
Catalog # HACHC20C310
Supplier:  Hach
CAS Number:  
Filling solutions for combined/reference electrodes
Filling solutions for combined/reference electrodes
KS120 KCl solution, saturated with AgCl, 500 ml bottle, 500 mL
CAS Number:  
Price
SFr. 91.30
Log in for availability
1 * 500 mL
Product Details & Documents
Filling and storage solutions for combined and reference electrodes.
Documents
Specifications
Specifications
  • Pk:
    500 mL
  • Description:
    KS120 KCl solution, saturated with AgCl, 500 ml bottle