Syringes, Digital Syringes™
22, 500 µl, 51 mm, 2, 750 RN
Catalog # 549-1296
Supplier: HAMILTON BONADUZ
Jump to:
Product Details & Documents
The Digital Syringe™ is a calibrated device composed of a serialised syringe and a serialised digital electronic holder.
- Achieve precision up to 0,5%
- Dispense volumes from 500 µl down to 50 nl
- Device also reduces the possibility of plunger bending
A digital readout allows enhanced syringe volume readability, while an adjustable plunger stop assures accurate, reproducible sample injections.
: Each unit is delivered with a certificate of calibration.
Specifications
- Gauge:22
- Type:750 RN
- Type of tip:2
- Volume:500 µl
- Length:51 mm
Accessories
Product Information
- Catalog #AvailabilityPrice
- Replacement battery for Digital Syringes™



