Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Stirring rods

Stirring rods

Fused and sealed at both ends, 8 mm, 300 mm
Catalog # 441-0149
Supplier:   Brand
Stirring rods
Catalog # 441-0149
Supplier:  Brand
Stirring rods
Stirring rods

Fused and sealed at both ends, 8 mm, 300 mm

Price
SFr. 80.70
Log in for availability
1 * 50 ST
Product Details & Documents

AR® glass, Glass tube is fused and sealed at both ends.

Specifications
  • Ø:
    8 mm
  • Description:
    Fused and sealed at both ends
  • Length:
    300 mm
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.