Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Sample vials in Lab File®, WHEATON®

Sample vials in Lab File®, WHEATON®

20,00 ml, 28×57 mm, Amber, No, 24-400
Catalog # WHEAW224815
Supplier:   DWK Life Sciences
Sample vials in Lab File®, WHEATON®
Catalog # WHEAW224815
Supplier:  DWK Life Sciences
Sample vials in Lab File®, WHEATON®
Sample vials in Lab File®, WHEATON®

20,00 ml, 28×57 mm, Amber, No, 24-400

Price
453.00 SFr.
Log in for availability
1 * 200 ST
Product Details & Documents

For storage of reagents and standards. Excellent for small derivatisation reactions. Wheaton 33® low extractable borosilicate glass is ideal for research studies where stability of glass is of prime importance.

Ordering information: Without cap.

Specifications
  • Cap included:
    No
  • Cap Size:
    24-400
  • Ø×H:
    28×57 mm
  • Capacity:
    20,00 ml
  • Colour:
    Amber
  • Seq:
    04