Nuts with double ferrules
Hex-head nut, short
Catalog # JOURJR-5511-5
Supplier: VICI JOUR RESEARCH
Jump to:
Product Details & Documents
These VICI Jour® nuts incorporate a unique PEEK double ferrule which grips the tubing in two places for added security, especially with PEEK or Tefzel tubing. For virtually any ¹/₁₆'' OD fittings eg. Parker, Swagelok, Waters, Valco, Rheodyne, Upchurch.
- Inert all-PEEK construction
- Fingertight and hex-head versions
- Rated up to 420 bar (6000 psi)
Specifications
- Description:Hex-head nut, short