Orders LinkContactUsLinkComponent
Forceps
Forceps
Fine point, straight, 120 mm
Catalog # 241-0338
Supplier:   EXCELTA
Forceps
Catalog # 241-0338
Supplier:  EXCELTA
Forceps
Fine point, straight, 120 mm
Price
84.20 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel, antimagnetic.
  • Economical special forceps for precision applications
  • Corrosion-resistant
Ideal when working with microelectronic assemblies and in the electronics industry.
Specifications
  • Description:
    Fine point, straight
  • Length:
    120 mm