Orders LinkContactUsLinkComponent
Face shields, Bionic™
Face shields, Bionic™
Anti-scratch, anti-fog, PC
Catalog # 111-0134
Supplier:   HONEYWELL SAFETY
Face shields, Bionic™
Catalog # 111-0134
Supplier:  HONEYWELL SAFETY
Face shields, Bionic™
Face shields, Bionic™
Anti-scratch, anti-fog, PC
Price
SFr. 93.50
Log in for availability
1 * 1 ST
Product Details & Documents
This innovative face shield provides protection from extreme mechanical hazards. It can be worn in combination with safety spectacles or respiratory protection.
  • With extended protection for brow and chin
  • Locking mechanism for added safety
  • Extra soft padding with washable or replaceable sweatband
  • Various adjustments for best comfort
Weight: 355 g.
EN 166, EN 169, EN 170, 1 F, B, T, C, 3, 8, 9
Certifications: EN 166, EN 169, EN 170, 1 F, B, T, C, 3, 8, 9
Specifications
  • Type:
    Anti-scratch, anti-fog
  • Material:
    PC
Accessories
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Replacement visor, uncoated
    Replacement visor, uncoated
    111-0740
    PC
    Log in for availability
    1 * 1 ST - SFr. 16.00
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Replacement visor, uncoated
    Replacement visor, uncoated
    111-2160
    Acetate
    Log in for availability
    1 * 1 ST - SFr. 26.10
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Replacement visor, anti-scratch/anti-fog
    Replacement visor, anti-scratch/anti-fog
    111-1293
    PC
    Log in for availability
    1 * 1 ST - SFr. 41.30
Product Information
  • Catalog #
    Availability
    Price
  • Replacement sweatband
    Replacement sweatband
    113-8053
    Log in for availability
    1 * 1 ST - SFr. 9.40
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..