Orders LinkContactUsLinkComponent
Lysates, human
Lysates, human
A431 whole cell lysate - EGF stimulated, 500 µG
Catalog # ROCKW09-000-362
Lysates, human
Catalog # ROCKW09-000-362
CAS Number:  
Lysates, human
Lysates, human
A431 whole cell lysate - EGF stimulated, 500 µG
CAS Number:  
Price
SFr. 325.00
Log in for availability
1 * 500 µG
Product Details & Documents
Whole cell lysates are derived from cell lines tissues using highly refined extraction protocols to ensure quality, protein integrity and high reproducibility. Lysates are prepared in denaturing buffer without dissociating agents.
  • Liquid
Whole cell lysates are ready to use for SDS-PAGE and Western blotting purposes. All extracts are tested by SDS-PAGE using 4 to 20% gradient gels and immunoblot analysis using antibodies to key cell signaling components to confirm the presence of both high molecular weight and low molecular weight proteins.
Each vial contains 1mg/ml of protein in SDS-PAGE sample buffer. The sample buffer consists of 0,05 mM Tris Hcl, 2% SDS, 10% glycerol and 0,1% bromophenol blue at pH 6,8.
Specifications
Specifications
  • Pk:
    500 µG
  • Description:
    A431 whole cell lysate - EGF stimulated