Orders LinkContactUsLinkComponent
Human plasma
Human plasma in sodium EDTA, sterile, 100 mL
  ROCKD519-04-0100
Human plasma
  ROCKD519-04-0100
 :  
Human plasma
Human plasma
Human plasma in sodium EDTA, sterile, 100 mL
 :  
SFr. 2’400.00
1 * 100 mL
Plasma is obtained from non-hemolysed blood collected from healthy, fasted donors. Sterilisation is accomplished by filtration.
  • Pk:
    100 mL
  • Description:
    Human plasma in sodium EDTA, sterile