Human plasma
Human plasma in sodium citrate, sterile, 1.000 mL
Catalog # ROCKD519-03-1000
Supplier: Rockland Immunochemicals
Jump to:
Product Details & Documents
Plasma is obtained from non-hemolysed blood collected from healthy, fasted donors. Sterilisation is accomplished by filtration.
Specifications
Specifications
- Pk:1.000 mL
- Description:Human plasma in sodium citrate, sterile