Human plasma
Human plasma in sodium citrate, sterile, 100 mL
ROCKD519-03-0100
Plasma is obtained from non-hemolysed blood collected from healthy, fasted donors. Sterilisation is accomplished by filtration.
Documents
D519-03-0050_ D519-04-0050_
Datasheet
- Pk:100 mL
- Description:Human plasma in sodium citrate, sterile