Human plasma
Human plasma in potassium EDTA, sterile, 500 mL
Catalog # ROCKD519-05-0500
Supplier: Rockland Immunochemicals
Jump to:
Product Details & Documents
Plasma is obtained from non-hemolysed blood collected from healthy, fasted donors. Sterilisation is accomplished by filtration.
Documents
D519-03-0050_ D519-04-0050_
Datasheet
Specifications
Specifications
- Pk:500 mL
- Description:Human plasma in potassium EDTA, sterile