Orders LinkContactUsLinkComponent
Human plasma
Human plasma
Human plasma in lithium heparin, sterile, 500 mL
Catalog # ROCKD519-07-0500
Human plasma
Catalog # ROCKD519-07-0500
CAS Number:  
Human plasma
Human plasma
Human plasma in lithium heparin, sterile, 500 mL
CAS Number:  
Price
SFr. 4’800.00
Log in for availability
1 * 500 mL
Product Details & Documents
Plasma is obtained from non-hemolysed blood collected from healthy, fasted donors. Sterilisation is accomplished by filtration.
Documents
Specifications
Specifications
  • Pk:
    500 mL
  • Description:
    Human plasma in lithium heparin, sterile