Orders LinkContactUsLinkComponent
Human plasma
Human plasma
Human plasma in lithium heparin, sterile, 50 mL
Catalog # ROCKD519-07-0050
Human plasma
Catalog # ROCKD519-07-0050
CAS Number:  
Human plasma
Human plasma
Human plasma in lithium heparin, sterile, 50 mL
CAS Number:  
Price
1 390.00 SFr.
Log in for availability
1 * 50 mL
Product Details & Documents
Plasma is obtained from non-hemolysed blood collected from healthy, fasted donors. Sterilisation is accomplished by filtration.
Documents
Specifications
Specifications
  • Pk:
    50 mL
  • Description:
    Human plasma in lithium heparin, sterile