Complement reagents
Rabbit C (2265 - 2720 g) complement
Katalognr. ROCKC204-0005
Direkt zu
Artikel gestrichen
Product Details & Documents
Complement reagents are prepared from healthy, non-medicated animals using very large pools to reduce lot-to-lot variability. The fresh frozen complement is rapidly processed and maintained at –70 °C to ensure maximum activity. The lyophilised complement is supplied with a diluent of veronal buffered saline containing optimal concentrations of magnesium and calcium.
Documents
C200-0005_ C201-0005_ C205-0005_ C220-0002_ C300-0050_
Fiche technique
Mehr
Technische Daten
- Description:Rabbit C (2265 - 2720 g) complement
- Seq:26
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



