Order Entry
Switzerland
Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class F2

VWR®, Calibration Weights, Class F2

2 kg, 30 ± mg
Catalog # 611-8168
Supplier:   VWR Collection
VWR®, Calibration Weights, Class F2
Catalog # 611-8168
Supplier:  VWR Collection
VWR®, Calibration Weights, Class F2
VWR®, Calibration Weights, Class F2

2 kg, 30 ± mg

Price
SFr. 238.00
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel Class F2 weights for users who require moderate levels of tolerance values in their calibration and day-to-day weighing.

  • Wire weights up to 500 mg in cobalt alloy
  • Identification number guarantees traceability for weights from 1 g upwards
  • Laser engraving improves legibility of the markings
  • Available with or without COFRAC certificate

Certifications: Conform to OILM R111 recommendations.

Ordering information: All weights from 1 mg to 20 kg are available with or without COFRAC certificate - please visit website or contact your local Avantor sales office. A range of sets of weights are also available - please search for product 'Calibration weight sets, Class F2'.

Packaging: Plastic cases have a vacuum sealed protective film to ensure that the case and calibration weight are protected during transportation.

Specifications
  • Weight:
    2 kg
  • Seq:
    54
  • Tolerance:
    30 ± mg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.