Order Entry
Switzerland
Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class F2

VWR®, Calibration Weights, Class F2

2 g, 0,4 ± mg
Catalog # 611-8159
Supplier:   VWR Collection
VWR®, Calibration Weights, Class F2
Catalog # 611-8159
Supplier:  VWR Collection
VWR®, Calibration Weights, Class F2
VWR®, Calibration Weights, Class F2

2 g, 0,4 ± mg

Price
40.50 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel Class F2 weights for users who require moderate levels of tolerance values in their calibration and day-to-day weighing.

  • Wire weights up to 500 mg in cobalt alloy
  • Identification number guarantees traceability for weights from 1 g upwards
  • Laser engraving improves legibility of the markings
  • Available with or without COFRAC certificate

Certifications: Conform to OILM R111 recommendations.

Informations de commande: All weights from 1 mg to 20 kg are available with or without COFRAC certificate - please visit website or contact your local Avantor sales office. A range of sets of weights are also available - please search for product 'Calibration weight sets, Class F2'.

Emballage: Plastic cases have a vacuum sealed protective film to ensure that the case and calibration weight are protected during transportation.

Specifications
  • Weight:
    2 g
  • Seq:
    45
  • Tolerance:
    0,4 ± mg