Fibronectin-coated flasks, Corning® BioCoat®
Cell culture flask, 250 ml, 75 cm², Plug seal screw cap, Canted
Supplier: Corning
Fibronectin-coated flasks, Corning® BioCoat®
Catalog # 734-0180
Supplier: Corning
Jump to:
Product Details & Documents
Human Fibronectin (HFN) is a widely distributed glycoprotein that is used as a substrate to promote attachment of cells through its central-binding domain RGD sequence. HFN is a product of most mesenchymal and epithelial cells and is present in both the ECM and plasma. The principal function of HFN appears to be in cellular migration during wound healing and development, regulation of cell growth and differentiation, and haemostasis/thrombosis
Applications include promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assays, studies of the effects of fibronectin on cell behaviour, and improving survival of primary cells in culture.
Documents
Certificates
Search by Lot NumberSpecifications
- Closure type:Plug seal screw cap
- Growth area:75 cm²
- Capacity:250 ml
- Description:Cell culture flask
- Seq:01
- Neck:Canted
- Cat. No.:734-0180
- Environmental certification:ISO 14001 - Bedford
- Environmentally Preferable:Low Manufacturing Impact



