Propionic acid ≥99%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 79-09-4 Molecular Formula: C3H6O2 Formula Weight: 74.08 Flash Point: 51°(123°F) Refractive Index: 1.3875 Odour: Unpleasant Melting Point: -21° Boiling Point: 140-141° Beilstein Reference: 506071 Merck Reference: 14,7825 UN No.: 3463 MDL No.: MFCD00002756
Synonyms: Propanoic acid
- CAS Number:79-09-4
- Formula:C₃H₆O₂
- MDL Number:MFCD00002756
- Molecular Weight:74.08 g/mol
- Merck Index:14,7825
- Boiling Pt:140…141 °C
- Melting Pt:–21 °C
- Density:0.992
- Flash Pt:51 °C (123 °F)
- Storage Temperature:Ambient
- EINECS:201-176-3
- UN:3463
- ADR:8,II
- REACH:01-2119486971-24
Documents
Certificates
Search by Lot NumberALFAL04210.22_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..



