Test kits, COD cell test, Spectroquant®, Supelco®
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*7 This method is officially recognised by the USEPA as an alternative method for the investigation of the following:
*7-1 wastewater
*7-2 drinking water
*7-3 respectively drinking water and wastewater
*8 COD cell tests fully comply with ISO 15705.
Documents
Specifications
Specifications
- Chemical method:Chromosulphuric oxidation
- Pk:25 Tests
- Description:COD cell test*⁷⁻¹
- For unit:Prove, Nova 30/60, Multy, Move
- Range:15 - 300 mg/L
- Cat. No.:1.14895.0001
- M-Clarity quality level:MQ100
Specifications
Accessories
Product Information
- Catalog #PkAvailabilityPrice
- Aborption cone for use with COD test for seawater 1.17059.0001Aborption cone for use with COD test for seawater 1.17059.00011.15955.00011Log in for availability1 * 1 ST - SFr. 218.00
Product Information
- Catalog #Pack typePkAvailabilityPrice