Cleanroom tapes
Cleanroom tape, 76,2 mm
817-0073
These tapes are made of polyolefin with acrylic adhesive.
- Ultra low outgassing
- Low sulfur, low halogen and low heavy metal
- Adhere lightly to plastics for easy removal
- Leaves no residue and particulates
Designed specifically for use inside semiconductor and critical environment cleanrooms. It contains extremely low leachable metals, eliminating the concern of contamination.
Length: 54,8 m
Colour: White
- Width:76,2 mm
- Description:Cleanroom tape



