Potassium hydroxide, pellets, EMSURE® for analysis, Supelco®
Fibre carton, 25 kg
Supplier: Merck
Potassium hydroxide, pellets, EMSURE® for analysis, Supelco®
Catalog # 1.05033.9025
Supplier: Merck
CAS Number: 1310-58-3
Jump to:
Product Details & Documents
Synonyms: Potash caustic, Caustic potash
- CAS Number:1310-58-3
- Formula:KOH
- MDL Number:MFCD00003553
- Molecular Weight:56.11 g/mol
- Merck Index:12,07806
- Boiling Pt:1320 °C (1013 hPa)
- Melting Pt:360 °C
- Density:2.04 g/cm³ (20 °C)
- EINECS:215-181-3
- ADR:8,II
- :M-Clarity™ quality level = MQ300
- Storage Temperature:Ambient
- UN:1813
- REACH:01-2119487136-33
Documents
Certificates
Search by Lot NumberMERC1.05033.0500_TDS MERC1.05033.1000_TDS_EN ProductPage_105033
Product specification
Merck Individual products and tailor-made chemicals for your technical applications
Product Literature
msds_105033
SDS
Specifications
Specification Test Results
- Solubility:1130 g/l (20 °C)
- Melting Point:360 °C
- Molar Mass:56.11 g/mol
- Boiling Point:1320 °C
- Vapor Pressure:(20 °C)
- Density:2.04 g/cm3 (20 °C)
- pH:14 (56 g/l, H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specifications
- Pack type:Fibre carton
- Pk:25 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



