Group Lock Boxes, Portable
Group lock box, mild sheet steel, for approx. 40 padlocks, Red, 227×89×152 mm
N° de catalogue BRDY065699
Fournisseur: Brady
Aller à
Product Details & Documents
For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps.
- Made of mild sheet steel
- Slot in front allows keys to be inserted when box is locked
Can also be used as a lock storage box holding up to 40 padlocks.
Documents
Spécifications
- Colour:Red
- Description:Group lock box, mild sheet steel, for approx. 40 padlocks
- Seq:02
- W×D×H:227×89×152 mm