Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Scoops

Scoops

37 ml, 105 mm, 30 mm
Catalog # NICK4022
Supplier:   NICKEL ELECTRO
Scoops
Catalog # NICK4022
Supplier:  NICKEL ELECTRO
Scoops
Scoops

37 ml, 105 mm, 30 mm

Price
SFr. 61.60
Log in for availability
1 * 5 ST
Product Details & Documents

Stainless steel. Round bottom. Ideal for use as weighing boats etc.

  • Excellent chemical resistance
  • Excellent thermal resistance
  • Smooth surfaces for fast, easy cleaning
Specifications
  • Width:
    30 mm
  • Capacity:
    37 ml
  • Length:
    105 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.